360 Degree Pre-labeled Solvent Resistant Spray Bottles, HCL Labels Inc
Supplier: HCL Label
BOT360SR-0002 BOT360SR-0003 BOT360SR-0005 BOT360SR-0006 BOT360SR-0007 BOT360SR-0008 BOT360SR-0009 BOT360SR-0010 BOT360SR-0011 BOT360SR-0012 BOT360SR-0013 BOT360SR-0014 BOT360SR-0016 BOT360SR-0017 BOT360SR-0018 BOT360SR-0019
103744-894EA 19.68 USD
103744-894 103744-896 103744-898 103744-900 103744-902 103744-904 103744-906 103744-908 103744-910 103744-912 103744-914 103744-916 103744-918 103744-920 103744-922 103744-924 103744-926 103744-928
360 Degree Pre-labeled Solvent Resistant Spray Bottles, HCL Labels Inc
Bottles Spray Bottles
HCL's pre-labeled GHS compliant 360 Degree solvent resistant spray bottles are durable, include a viton reinforced O-ring, and are affixed with a laminated adhesive vinyl GHS label for additional chemical and solvent resistance.
  • 360 Degree sprayer can be continuously sprayed sideways or upside-down, with no leakage
  • Labels are made of a durable adhesive vinyl that is laminated for chemical and solvent resistance
HCL's GHS compliant 360 Degree solvent resistant spray bottles by Birchmeier are pre-affixed with a chosen GHS hazard communication label. These bottles include a special viton seal to prevent leakage, and are supplied empty and ready to be filled.