Nitrile Extended-Cuff, Chemo-Rated, Powder-Free, Fingertip-Textured Exam Gloves, Tronex®
Supplier: Tronex
9099-10 9099-20 9099-30 9099-35
75923-408BX 29.54 USD
75923-408 75923-410 75923-412 75923-414 75923-416
Nitrile Extended-Cuff, Chemo-Rated, Powder-Free, Fingertip-Textured Exam Gloves, Tronex®
Gloves
Nitrile gloves offer exam-grade protection with an extended cuff, fingertip texturing, and they are chemo-rated.
  • Exclusive material formulation with 12" cuff and added material thickness for extended coverage
  • Chemo rated per ASTM D6978
  • Not made with natural rubber latex
  • Powder-free and nonsterile
  • Fingertip textured for improved grip
Exclusive nitrile material formulation offering enhanced durability and abrasion resistance, while featuring an extended cuff for expanded coverage area. Provides excellent barrier protection from bloodborne pathogens, as well as exceptional resistance to chemotherapy drugs and other chemicals. These gloves provide a high degree of tensile strength and elongation combined with comfort, flexibility, and dexterity.
Envase: Gloves are packaged 100 per box, 10 boxes per case.