Uvex Stealth® Reader Goggles, Honeywell Safety

Supplier: Honeywell Safety Products
S3992X S3993X S3991X
89140-320EA 59.62 USD
89140-320 89140-322 89140-318
Uvex Stealth® Reader Goggles, Honeywell Safety
Goggles
These reader goggles are made of soft elastomer body which conforms to facial contours for a secure, customized, gap-free fit. Ideal for protection as well as increased magnification for close-up inspection or fine detail work.

  • Uvex Stealth® OTG lenses used for goggle body
  • Economical lens replacement system extends the life of the goggle
  • Protect against hazards like abrasion, airborne particulates, biohazards, chemicals and splash
  • Lens thickness 0.10" (2.55 mm)
  • Lens overall width 6.81" (173 mm)

The low profile, wrap-around style provides complete coverage and unobstructed vision and protection from chemical splash. The wide, adjustable slide headband with pivoting clips provides an easy-to-adjust fit for complete comfort, even with hard hats.

Certificaciones: Meets ANSI Z87+ - Meets ANSI Z87+ (High Impact) standard, CSA Z94.3

Envase: Box contains 50/Case (Goggles) and 50/Case Ind. Polybag, 10/Bag, 200/Box (Replacement Lenses).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR