Zinc D-gluconate hydrate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Gluconic acid Zzinc(II) salt hydrate, Zinc(II) gluconate hydrate, Zinc gluconate hydrate

ZI108-2.5KG ZI108-500GM ZI108-12KG ZI108-50KGBL
75873-594EA 407.05 USD
75873-594 75873-596 76015-020 76015-022
Zinc D-gluconate hydrate
Zinc D-gluconate hydrate

Zinc Gluconate, Hydrate is the zinc salt of gluconic acid can also be manufactured by fermenting glucose. It comes as a white powder as is used in cold treating lozenges and as one of the more effective ways of delivering zinc as a dietary supplement.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₂₂O₁₄Zn·xH₂O
MW: 455.69 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00868110
CAS Number: 4468-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR