Zymosan A

Supplier: Spectrum Chemical Mfg. Corp.

Z3003-1GM Z3003-500GMBL Z3003-5GM
75873-580EA 541.35 USD
75873-580 75873-582 75873-584
Zymosan A
Zymosan A
Storage Temperature: Refrigerator
MDL Number: MFCD00082157
CAS Number: 58856-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR