p-Xylylenediamine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,4-Bis(aminomethyl)benzene, 1,4-Phenylenedimethanamine

X1022-100GM X1022-25GM
75873-562EA 401.6 USD
75873-562 75873-564
p-Xylylenediamine
p-Xylylenediamine
Formula: C₈H₁₂N₂
MW: 136.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009821
CAS Number: 539-48-0
UN: 3259
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR