Tetrabutylammonium hexafluorophosphate

Supplier: SPECTRUM CHEMICAL MFG CORP

T2646-250GM T2646-25GM
75873-492EA 351.75 USD
75873-492 75873-494
Tetrabutylammonium hexafluorophosphate
Tetrabutylammonium hexafluorophosphate
Formula: C₁₆H₃₆F₆NP
MW: 387.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011748
CAS Number: 3109-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR