2,2,2-Trichloroethanol

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2,2,2-Trichloroethan-1-ol

T2628-100GM T2628-500GM
75873-488EA 65.14 USD
75873-488 75873-490
2,2,2-Trichloroethanol
2,2,2-Trichloroethanol

2,2,2-Trichloroethanol is related to ethanol and can be used as a sedative or a hypnotic.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂H₃Cl₃O
MW: 149.4 g/mol
Density: 1.557
Storage Temperature: Ambient
MDL Number: MFCD00004677
CAS Number: 115-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR