Tetrakis(triphenylphosphine)palladium(0)

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4

T2020-1GM T2020-200GMBL T2020-25GM T2020-5GM
75873-464EA 96.66 USD
75873-464 75873-466 75873-468 75873-470
Tetrakis(triphenylphosphine)palladium(0)
Tetrakis(triphenylphosphine)palladium(0)
Formula: C₇₂H₆₀P₄Pd
MW: 1155.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010012
CAS Number: 14221-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR