Tetrabutylammonium tetrafluoroborate
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Tetrabutylammoniumtetrafluoroborate
T1692-100GM
T1692-25GM
75873-460EA
308
USD
75873-460
75873-462
Tetrabutylammonium tetrafluoroborate
Tetrabutylammonium tetrafluoroborate
Formula:
C₁₆H₃₆BF₄N MW: 329.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011634 CAS Number: 429-42-5 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...