o-Toluidinium chloride

Supplier: Spectrum Chemical Mfg. Corp.

T1680-100GM T1680-25GM
75873-444EA 369.05 USD
75873-444 75873-446
o-Toluidinium chloride
o-Toluidinium chloride
Formula: C₇H₁₀ClN
Storage Temperature: Ambient
MDL Number: MFCD00012959
CAS Number: 636-21-5
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR