Terephthalic acid dihydrazide

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Terephthalic Dihydrazide

T1552-100GM T1552-25GM
75873-404EA 283.55 USD
75873-404 75873-406
Terephthalic acid dihydrazide
Terephthalic acid dihydrazide
Formula: C₈H₁₀N₄O₂
MW: 194.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014762
CAS Number: 136-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR