Terephthalonitrile

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,4-Benzenedicarbonitrile, Terephthalodinitrile, Benzene-1,4-dicarbonitrile, 1,4-Dicyanobenzene

T1518-100GM T1518-25GM
75873-400EA 355.45 USD
75873-400 75873-402
Terephthalonitrile
Terephthalonitrile
Formula: C₈H₄N₂
MW: 128.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001810
CAS Number: 623-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR