Tetrabutylammonium hydrogen sulfate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate

T1294-100GM T1294-25GM T1294-5GM
75873-370EA 549.52 USD
75873-370 75873-372 75873-374
Tetrabutylammonium hydrogen sulfate
Tetrabutylammonium hydrogen sulphate

Tetrabutylammonium Hydrogen Sulfate is a chemical compound and is most often employed as a solution in alcohols or water. Tetrabutylammonium hydrogen sulfate is an ion pairing reagent that is used in liquid chromatography. It is frequently utilized in the analysis of pharmaceutical compounds and metabolites.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: (C₄H₉)₄NHSO₄
MW: 339.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011637
CAS Number: 32503-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR