Suberic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,8-Octanedioic acid, Octanedioic acid, 1,6-Hexanedicarboxylic acid

S2023-100GM S2023-500GM S2023-5GM S2023-50KGBL
75873-234EA 233.2 USD
75873-234 75873-278 75873-280 76014-904
Suberic acid
Suberic acid

Suberic Acid, also known as octanedioic acid, is a dicarboxylic acid used in plastics manufacture and drug syntheses.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004428
CAS Number: 505-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR