L(-)-Proline

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH

PR120-100GM PR120-1KG PR120-25GM PR120-5KG
75873-274EA 147.1 USD
75873-274 75873-276 75873-284 75873-286
L(-)-Proline
L(-)-Proline

L-Proline, also known simply as proline, is a DNA-encoded amino acids and can be used as a asymmetric catalyst in organic reactions.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 245
Storage Temperature: Ambient
MDL Number: MFCD00064318
CAS Number: 147-85-3
Merck Index: 13,07871

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR