L-Phenylalanine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine

PH145-100GM PH145-1KG PH145-25GM
75873-268EA 132.39 USD
75873-268 75873-270 75873-272
L-Phenylalanine
L-Phenylalanine

L-Phenylalanine, also known as 2-Amino-3-phenylpropanoic acid, is an essential amino acid used to biochemically form proteins that are coded for by DNA.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR