1-Pentadecanol
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Pentadecan-1-ol, 1-Pentadecanol
P2358-100GM
P2358-25GM
75873-248EA
875.15
USD
75873-248
75873-250
1-Pentadecanol
1-Pentadecanol
1-Pentadecanol is a fatty alcohol and used in organic synthesis.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
H₃C(CH₂)₁₃CH₂OH MW: 228.42 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004759 CAS Number: 629-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...