4-Picolyl chloride hydrochloride
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
4-(Chloromethyl)pyridine hydrochloride, 4-(Chloromethyl)-pyridinium chloride
P2064-100GM
P2064-25GM
75873-204EA
500.65
USD
75873-204
75873-206
4-Picolyl chloride hydrochloride
4-Picolyl chloride hydrochloride
Formula:
C₆H₇Cl₂N MW: 164.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012826 CAS Number: 1822-51-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...