Dipotassium thiosulfate

Supplier: Spectrum Chemical Mfg. Corp.

P1718-100GM P1718-25GM P1718-500GM
75873-194EA 535.9 USD
75873-194 75873-196 75873-198
Dipotassium thiosulfate
Dipotassium thiosulphate
Formula: K₂O₃S₂
Storage Temperature: Ambient
MDL Number: MFCD00036292
CAS Number: 10294-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR