Phenyl-4-hydroxybenzoate

Supplier: SPECTRUM CHEMICAL MFG CORP

P1504-100GM P1504-25GM
75872-782EA 462.65 USD
75872-782 75872-784
Phenyl-4-hydroxybenzoate
Phenyl 4-hydroxybenzoate

Phenyl 4-Hydroxybenzoate is used as a cosmetic preservative.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₃H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00016467
CAS Number: 17696-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR