Potassium dihydrogen citrate

Supplier: Spectrum Chemical Mfg. Corp.

P1268-2KG P1268-500GM
75873-138EA 502.05 USD
75873-138 75873-140
Potassium dihydrogen citrate
Potassium dihydrogen citrate

Potassium Citrate Monobasic is a white, crystalline powder that is hygroscopic. It has no odor and has a slightly salty taste. It is a potassium salt of citric acid and is most frequently used as a buffering agent, sequestrant and acidity regulator.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₇KO₇
Storage Temperature: Ambient
MDL Number: MFCD00036472
CAS Number: 866-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR