Potassium acetate, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Acetic acid potassium salt

P1188-2.5KG P1188-500GM
76014-736EA 1607.75 USD
76014-736 76014-738
Potassium acetate, BiotechGrade
Potassium acetate

Potassium Acetate, BiotechGrade is used in molecular biology to precipitate dodecyl sulfate (DS) and DS-bound proteins, allowing the removal of proteins from DNA. It is also used as a salt for the ethanol precipitation of DNA. Potassium acetate is also used in mixtures applied for tissue preservation, fixation, and mummification.

Formula: CH₃COOK
MW: 98.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR