1-Naphthylacetic acid
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Planofix, 1-Naphthaleneacetic acid
NA107-100GM
NA107-1KG
NA107-25GM
NA107-500GM
NA107-5KG
NA107-25KGBL
75873-104EA
127.13
USD
75873-104
75873-106
75873-108
75873-110
75873-112
76014-706
1-Naphthylacetic acid
1-Naphthylacetic acid
1-Naphthaleneacetic Acid is an organic compound that is colorless and soluble in organic solvents. It is a synthetic plant hormone that is part of the auxin family and is found in most commercial horticultural plant rooting products and is used for rooting and propagation purposes
Formula:
C₁₂H₁₀O₂ MW: 186.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004046 CAS Number: 86-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...