(±)-Nipecotic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: H-DL-Nip-OH, (±)-3-Piperidinecarboxylic acid, Piperidine-3-carboxylic acid, Nipecotic acid

N3821-100GM N3821-25GM
75873-100EA 310.7 USD
75873-100 75873-102
(±)-Nipecotic acid
(±)-Nipecotic acid
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005992
CAS Number: 498-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR