2-Nitrobenzaldehyde

Supplier: SPECTRUM CHEMICAL MFG CORP

N2155-100GM N2155-25GM
75872-436EA 287.89 USD
75872-436 75872-438
2-Nitrobenzaldehyde
2-Nitrobenzaldehyde

2-Nitrobenzaldehyde, also known as Nitrobenzaldehyde or ortho-nitrobenzaldehyde, is an organic aromatic compound used as an intermediate to synthesize the dye Indigo.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00007132
CAS Number: 552-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR