4'-Nitroacetanilide

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: N-(4-Nitrophenyl)acetamide

N1522-100GM N1522-25GM
75873-060EA 260.5 USD
75873-060 75873-062
4'-Nitroacetanilide
4'-Nitroacetanilide
Formula: C₈H₈N₂O₃
MW: 180.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007303
CAS Number: 104-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR