1-Nitro-9,10-anthraquinone

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1-Nitroanthraquinone, 1-Nitroanthracene-9,10-dione

N1512-100GM N1512-25GM
75873-050EA 325.65 USD
75873-050 75873-052
1-Nitro-9,10-anthraquinone
1-Nitro-9,10-anthraquinone
Formula: C₁₄H₇NO₄
MW: 253.2097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019140
CAS Number: 82-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR