Methacrylic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Methylpropenoic acid

M2131-100ML M2131-500MLGL
75873-004EA 94.35 USD
75873-004 75873-006
Methacrylic acid
Methacrylic acid

Methacrylic Acid is used as a building block to make a wide range of polymer-based products like binders in paints and coatings, water-soluble polymers and interlevant in glass.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₆O₂
MW: 86.09 g/mol
Density: 1.015
Storage Temperature: Ambient
MDL Number: MFCD00002651
CAS Number: 79-41-4
UN: 2531
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR