(R)-(-)-Mandelic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid

M1584-100GM M1584-10KGBL M1584-1KGBL M1584-25GM
75872-940EA 257.75 USD
75872-940 75872-942 75872-944 75872-946
(R)-(-)-Mandelic acid
(R)-(-)-Mandelic acid

R-(-)-Mandelic Acid, also known as Phenylglycolic acid, is an aromatic alpha hydroxy acid.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₈H₈O₃
MW: 152.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064251
CAS Number: 611-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR