L(+)-Leucine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine

LE115-100GM LE115-1KG LE115-250GM LE115-25GM LE115-5KG
75872-884EA 108.22 USD
75872-884 75872-886 75872-888 75872-890 75872-892
L(+)-Leucine
L(+)-Leucine

L-Leucine, also known as 2-Amino-4-methylpentanoic acid, is a component of the subunits in astacin, ferritin, and other buffer proteins.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002617
CAS Number: 61-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR