4-Methylquinoline

Supplier: SPECTRUM CHEMICAL MFG CORP

L1606-100ML L1606-25ML
75872-856EA 478.95 USD
75872-856 75872-858
4-Methylquinoline
4-Methylquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006784
CAS Number: 491-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR