N-Lauroylsarcosine sodium salt

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Lauroylsarcosine sodium, Sarkosyl, Sodium lauroyl sarcosine, Sodium N-lauroylsarcosinate, Sodium N-dodecanoyl-N-methylglycinate

L1228-100GM L1228-2.5KG L1228-25GM L1228-500GM L1228-12KGBL
75872-838EA 122.93 USD
75872-838 75872-840 75872-842 75872-844 76014-588
N-Lauroylsarcosine sodium salt
N-Lauroylsarcosine sodium salt
Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MW: 293.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042728
CAS Number: 137-16-6
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0126d80345cce5dd675a83796c379bd44cebacef&vid=48c74917d6a93a32467317ef6c6fdbf24f3b6dc2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR