Iodine solution 5% (w/v) USP, strong (DEA List I Chemical)
Supplier: SPECTRUM CHEMICAL MFG CORP
IO106-100ML
IO106-200LTBL
IO106-20LTBL
IO106-4LTGL
IO106-4LTGL-CS
IO106-500ML-CS
IO106-500MLGL
76014-552EA
131.2
USD
76014-552
76014-554
76014-556
76014-558
76014-560
76014-562
76014-564
Iodine solution 5% (w/v) USP, strong (DEA List I Chemical)
Iodine
Strong Iodine Solution, 5 Percent (w/v) (DEA List I Chemical), USP.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
I₂ Density: 1.007 Storage Temperature: Ambient |
MDL Number:
MFCD00011355 CAS Number: 7553-56-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...