3-Hydroxy-p-anisaldehyde
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde
I3248-100GM
I3248-25GM
75872-822EA
297.1
USD
75872-822
75872-824
3-Hydroxy-p-anisaldehyde
3-Hydroxy-p-anisaldehyde
Formula:
C₈H₈O₃ MW: 152.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003369 CAS Number: 621-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...