Indanthrone

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Dinaphtho[2,3-a:2',3'-h]phenazine-5,9,14,18(6H,15H)-tetraone

I1276-100GM I1276-25GM
75872-742EA 264.6 USD
75872-742 75872-750
Indanthrone
Indanthrone
Formula: C₂₈H₁₄N₂O₄
MW: 442.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046964
CAS Number: 81-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR