N-Isopropylcyclohexylamine

Supplier: Spectrum Chemical Mfg. Corp.

I1270-100ML I1270-25ML
75872-736EA 331.1 USD
75872-736 75872-738
N-Isopropylcyclohexylamine
N-Isopropylcyclohexylamine
Formula: C₉H₁₉N
Storage Temperature: Ambient
MDL Number: MFCD00003831
CAS Number: 1195-42-2
UN: 2924
ADR: 38,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR