1-Hexadecene

Supplier: Spectrum Chemical Mfg. Corp.

H3297-100ML H3297-25ML
75872-704EA 229.3 USD
75872-704 75872-706
1-Hexadecene
1-Hexadecene
Formula: C₁₆H₃₂
Density: 0,783
Storage Temperature: Ambient
MDL Number: MFCD00008991
CAS Number: 629-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR