1-Heptene
Supplier: SPECTRUM CHEMICAL MFG CORP
H2666-100ML
H2666-25ML
75872-688EA
351.4
USD
75872-688
75872-690
1-Heptene
1-Heptene
Formula:
C₇H₁₄ MW: 98.19 g/mol Density: 0.697 Storage Temperature: Ambient |
MDL Number:
MFCD00009531 CAS Number: 592-76-7 UN: 2278 ADR: 3,II |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...