N-Hydroxysuccinimide

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu

H2495-100GM H2495-250GM H2495-25GM H2495-25KGBL
75872-682EA 177.57 USD
75872-682 75872-684 75872-686 76014-530
N-Hydroxysuccinimide
N-Hydroxysuccinimide

N-Hydroxysuccinimide is an organic compound often found in biochemistry and used as an activating reagent for carboxylic acids.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₅NO₃
MW: 115.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005516
CAS Number: 6066-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR