Hydrindantin dihydrate

Supplier: Spectrum Chemical Mfg. Corp.

H2451-100GM H2451-25GM H2451-5GM
75872-676EA 2279.3 USD
75872-676 75872-678 75872-680
Hydrindantin dihydrate
Hydrindantin dihydrate
Formula: C₁₈H₁₀O₆·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149242
CAS Number: 5950-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR