Heptyl-4-hydroxybenzoate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Heptylparaben, n-Heptyl 4-hydroxybenzoate, Heptyl parahydroxybenzoate, 4-Hydroxybenzoic acid heptyl ester

H1532-100GM H1532-25GM
75872-660EA 276.8 USD
75872-660 75872-662
Heptyl-4-hydroxybenzoate
Heptyl-4-hydroxybenzoate
Formula: C₁₄H₂₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00016481
CAS Number: 1085-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR