2-Hydroxyethyl acetate

Supplier: SPECTRUM CHEMICAL MFG CORP

H1466-100ML H1466-25ML
75872-618EA 317.45 USD
75872-618 75872-620
2-Hydroxyethyl acetate
2-Hydroxyethyl acetate
Formula: C₄H₈O₃
MW: 104.1045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002864
CAS Number: 542-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR