Hexyl-p-toluenesulfonate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: p-Toluenesulphonic acid Hexyl Ester, Hexyl p-Toluenesulphonate

H1463-100GM H1463-25GM
75872-422EA 257.75 USD
75872-422 75872-616
Hexyl-p-toluenesulfonate
Hexyl-p-toluenesulfonate
Formula: C₁₃H₂₀O₃S
MW: 256.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059439
CAS Number: 3839-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR