L(+)-Glutamine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q

GL136-100GMGL GL136-1KGGL GL136-25GMGL
75872-590EA 149.2 USD
75872-590 75872-592 75872-594
L(+)-Glutamine
L(+)-Glutamine

L-Glutamine is also known as glutamine and is a free amino acid in the human blood.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS Number: 56-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=15d7d87daff0fec33a974a578415e157a3278049&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR