Sodium-L(+)-glutamate monohydrate

Supplier: SPECTRUM CHEMICAL MFG CORP

GL135-2.5KG GL135-500GM GL135-12KG
75872-586EA 145.25 USD
75872-586 75872-588 76014-518
Sodium-L(+)-glutamate monohydrate
Sodium-L(+)-glutamate monohydrate

L-Glutamic Acid Monosodium Salt is also known as sodium glutamate and is used in the food industry most usually as a flavorant.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅H₈NNaO₄·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150138
CAS Number: 6106-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR