Glucosamine sulfate potassium chloride USP dietary supplement
Supplier: SPECTRUM CHEMICAL MFG CORP
G1296-100GM
G1296-12KG
G1296-2.5KG
G1296-25KGBL
G1296-500GM
76014-506EA
138.4
USD
76014-506
76014-508
76014-510
76014-512
76014-514
Glucosamine sulfate potassium chloride USP dietary supplement
Glucosamine sulfate potassium chloride
Glucosamine Sulfate Potassium Chloride, Dietary Supplement, USP.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
(C₆H₁₄NO₅)₂SO₄·2KCl MW: 605.52 g/mol Storage Temperature: Ambient |
CAS Number:
1296149-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...