D-(+)-Glucosamine hydrochloride USP dietary supplement

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Amino-2-deoxy-D-glucopyranose Hydrochloride, D-Glucosamine hydrochloride, (2R,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride

G1094-100GM G1094-1KG G1094-25KGBL G1094-500GM G1094-5KG
76014-494EA 139.73 USD
76014-494 76014-496 76014-498 76014-500 76014-502
D-(+)-Glucosamine hydrochloride USP dietary supplement
D-(+)-Glucosamine hydrochloride

Glucosamine Hydrochloride, USP Dietary Supplement is a dietary supplement with osteoarthritis pain relief properties.


All Spectrum Chemical USP Dietary Supplement grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Formula: C₆H₁₃NO₅
MW: 215.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00135831
CAS Number: 66-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR