Glycolic acid 70%, high purity

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Hydroxy acid, 2-Hydroxyacetic acid

G1059-100ML G1059-200LTBL G1059-20LT G1059-4LTGL G1059-4LTGL-CS G1059-500ML-CS G1059-500MLGL
76014-480EA 111.5 USD
76014-480 76014-482 76014-484 76014-486 76014-488 76014-490 76014-492
Glycolic acid 70%, high purity
Glycolic acid

Glycolic Acid, 70 Percent, High Purity, is slightly stronger than acetic acid. It is often included into emulsion polymers, solvents and additives for ink and paint in order to improve flow properties and impart gloss. It is used in surface treatment products that increase the coefficient of friction on tile flooring. Glycolic acid is a useful synthetic organic intermediate for oxidation-reduction, esterification and long chain polymerizations

Formula: C₂H₄O₃
MW: 76.05 g/mol
Density: 1.25
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS Number: 79-14-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR