Fluorescein disodium salt

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt, Acid Yellow 73, Yellow No. 8, Fluorescein sodium salt, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate

F1272-100GM F1272-25GM F1272-500GM
75872-508EA 93.51 USD
75872-508 75872-510 75872-512
Fluorescein disodium salt
Fluorescein disodium salt

Fluorescein Sodium is a synthetic organic compound that is a common fluochrome dye. It is obtained from the condensation of phthalic anhydride with resorcinol on heating. It is used in dying silk and wool, ink manufacturing, in coloring cosmetics, and in coloring gasoline and as a toner. It is widely used as a fluorescent tracer for many applications. It appears as a yellowish (yellowish-red) to red solid crystalline powder that is soluble in acetone, methanol, hot alcohol, glacial acetic acid, alkali hydroxides, carbonates and water.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₁₀Na₂O₅
MW: 376.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00167039
CAS Number: 518-47-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR